Learn More
Sino Biological Dengue virus (DENV) (type 2, strain New Guinea C) NS1 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40263T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C) NS1 ( Catalog#40263-V07H; AAC59275.1; Asp1-Ala352). Dengue virus (DENV) (type 2, strain New Guinea C) NS1 specific IgG was purified by Dengue virus (DENV) (type 2, strain New Guinea C) NS1 affinity chromatography.
Catalog No. 50-202-2607