Learn More
Sino Biological Dengue virus (DENV) (type 2, strain New Guinea C) NS1 Protein (His Tag)
Supplier:  Sino Biological 40263V07H100
Encompass_Preferred
A DNA sequence encoding the Dengue virus (type 2, strain New Guinea C) NS1 (AAC59275.1) (Asp776-Ala1127) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0680