Learn More
Sino Biological Hepatitis C virus (HCV-1a) NS3 protease / helicase immunodominant region Protein (aa 1356-1459, GST Tag)
Catalog No. 501620686
Encompass_Preferred
A DNA sequence encoding the Hepatitis C virus (HCV)(serotype 1a, isolate H77) NS3 (NP_803144.1) (Thr1356-Thr1459) was expressed with a GST tag at the N-terminus.
Catalog No. 50-162-0686 Supplier Sino Biological Supplier No. 40286V09E100
A DNA sequence encoding the Hepatitis C virus (HCV)(serotype 1a, isolate H77) NS3 (NP_803144.1) (Thr1356-Thr1459) was expressed with a GST tag at the N-terminus.