Learn More
Sino Biological Recombinant P. falciparum pfHPRT Protein (His Tag)
Supplier:  Sino Biological 40301N07E100
Encompass_Preferred
A DNA sequence encoding the pfHPRT (CZT98374.1) (Met1-Leu231) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0689