Learn More
Sino Biological Recombinant P. falciparum PfLDH Protein (His Tag)
Supplier:  Sino Biological 40303N08E100
Encompass_Preferred
A DNA sequence encoding the PfLDH (XP_001349989.1) (Met1-Ala316) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0691