Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain Mayinga 1976) Glycoprotein / GP-RBD (Receptor Binding Domain) Protein (Fc Tag)
Supplier:  Sino Biological 40304V02H1100
Encompass_Preferred
A DNA sequence encoding the Zaire ebolavirus (strain Mayinga 1976) GP (AAC54887.1) (Met1-Phe308) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0693