Learn More
Sino Biological Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40323V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/little yellow-shouldered bat/Guatemala/164/2009 (H17N10)) hemagglutinin (AFC35418.1) (Met1-Tyr527), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0701