Learn More
Sino Biological Influenza A H7N9 (A/Zhejiang/DTID-ZJU10/2013) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40325V08B100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Zhejiang/DTID-ZJU10/2013(H7N9)) hemagglutinin (AHA11500.1) (Met1-Val524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0710