Learn More
eMolecules​ 4033-39-0 | L-2,3-Diaminopropionic acid | AA Blocks LLC | MFCD09265268 | 104.109 | C3H8N2O2 | 0.000 | NC[C@H](N)C(O)=O | 250mg | 913102600
Catalog No. NC3966786 Shop All eMolecules Inc. Products
Encompass_Preferred
L-2,3-Diaminopropionic acid | AA Blocks LLC | 4033-39-0 | MFCD09265268 | 104.109 | C3H8N2O2 | 0.000 | NC[C@H](N)C(O)=O | 250mg | 913102600
Catalog No. NC3966786 Supplier eMolecules​ Supplier No. 913102600
L-2,3-Diaminopropionic acid | AA Blocks LLC | 4033-39-0 | MFCD09265268 | 104.109 | C3H8N2O2 | 0.000 | NC[C@H](N)C(O)=O | 250mg | 913102600