Learn More
Sino Biological Hendra virus (HeV) (isolate Horse/Autralia/Hendra/1994) Glycoprotein Protein (Fc Tag)
Supplier:  Sino Biological 40335V02H100
Encompass_Preferred
A DNA sequence encoding glycoprotein G of the Hendra virus (isolate Horse/Autralia/Hendra/1994) (AAC83193.2) (Gln71-Ser604) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0715