Learn More
Sino Biological Rift Valley fever virus (RVFV) (strain MP12) glycoprotein / G2 Protein (His Tag)
Supplier:  Sino Biological 40338V07B1100
Encompass_Preferred
A DNA sequence encoding the Rift Valley fever virus (RVFV) (strain MP12) Gc (ABD38821.1) (Cys691-Ser1139) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0718