Learn More
Sino Biological West Nile Virus (WNV) (lineage 1, strain NY99) E / Envelope Protein (Domain III, His Tag)
Supplier:  Sino Biological 40345V08Y100
Encompass_Preferred
A DNA sequence encoding the domain III of the West Nile virus (WNV)((lineage 1, strain NY99) envelope glycoprotein (E) (ABA62343.1) (Gln586-Ile694) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0723