Learn More
Sino Biological West Nile Virus (WNV) (lineage 1, strain NY99) NS1 Protein (His Tag)
Supplier:  Sino Biological 40346V07H100
Encompass_Preferred
A DNA sequence encoding the NS1 of West Nile Virus (WNV)(lineage 1, strain NY99) (YP_001527881.1) (Asp1-Ala352) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0727