Learn More
Sino Biological Influenza A H1N1 (A/California/06/2009) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40350V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/California/06/2009(H1N1)) hemagglutinin (ACP41935.1)(Met1-Arg344), termed as HA1,was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0731