Learn More
Sino Biological Influenza A H10N8 (A/duck/Guangdong/E1/2012) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40351V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Guangdong/E1/2012(H10N8)) hemagglutinin (Met1-Arg340), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0733