Learn More
Sino Biological Influenza A H10N8 (A/duck/Guangdong/E1/2012) Neuraminidase / NA Protein (His Tag)
Supplier:  Sino Biological 40352V07B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Guangdong/E1/2012(H10N8)) Neuraminidase (AFJ19054.1) (Gly38-Gly470) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0735