Learn More
Sino Biological Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40359V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Jiangxi-Donghu/346/2013(H10N8)) hemagglutinin (EPI497477) (Met1-Asp524) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0741