Learn More
Sino Biological Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin / HA0 Protein (full length)
Supplier:  Sino Biological 40359VNAB100
Encompass_Preferred
A DNA sequence encoding the full length of Influenza A virus (A/Jiangxi-Donghu/346/2013(H10N8)) hemagglutinin (translated amino acid sequence of EPI497477 ) (Met1-Ile561),termed as HA, was expressed.
Catalog No. 50-162-0746