Learn More
Sino Biological Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) Glycoprotein / GP-RBD (Receptor Binding Domain) Protein (Fc Tag)
Supplier:  Sino Biological 40368V02H100
Encompass_Preferred
A DNA sequence encoding the Bundibugyo ebolavirus(strain Uganda 2007) GP (ACI28624.1) (Met1-Val308) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0750