Learn More
Sino Biological Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) GP-RBD / Glycoprotein Protein (Fc Tag)
Catalog No. 501620751
Encompass_Preferred
A DNA sequence encoding the Bundibugyo ebolavirus(strain Uganda 2007) GP (ACI28624.1) (Arg54-Glu201) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0751 Supplier Sino Biological Supplier No. 40368V02H1100
A DNA sequence encoding the Bundibugyo ebolavirus(strain Uganda 2007) GP (ACI28624.1) (Arg54-Glu201) was expressed with the Fc region of human IgG1 at the C-terminus.