Learn More
Sino Biological Macaca fascicularis Cytomegalovirus (CyCMV) (strain OT-1) glycoprotein B / GB Protein (Fc Tag)
Supplier:  Sino Biological 40369V02H100
Encompass_Preferred
A DNA sequence encoding the macaca fascicularis cytomegalovirus (CyCMV)(strain OT-1) glycoprotein B(gB) (ADV35145.1) (Met1-Asp680, with cleavage site mutated ‘RHKR’ to ‘TTQT’) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0756