Learn More
Sino Biological Influenza A H5N1 (A/chicken/Jilin/9/2004) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40372V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/chicken/Jilin/9/2004(H5N1)) hemagglutinin (Met1-Arg342), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0762