Learn More
Sino Biological Epstein-Barr virus (Herpesvirus 4) EBV Glycoprotein gp350 / EBV GP350 Protein (His Tag)
Supplier:  Sino Biological 40373V08B100
Encompass_Preferred
A DNA sequence encoding the human herpesvirus 4/Epstein-Barr virus gp350 (strain B95-8) (YP_401667.1) (Met1-Pro490) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0764