Learn More
Sino Biological Hepatitis C virus Envelope Glycoprotein E1 / HCV-E1 (subtype 1b, strain HC-J4) Protein (His Tag)
Supplier:  Sino Biological 40374V07H100
Encompass_Preferred
A DNA sequence encoding the envelope glycoprotein E1 of Hepatitis C virus (subtype 1b, strain HC-J4) (AAC15725.1) (Tyr192-Ile340) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0769