Learn More
Sino Biological Influenza A H5N8 (A/breeder duck/Korea/Gochang1/2014) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40383V08H2100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/breeder duck/Korea/Gochang1/2014 (H5N8)) hemagglutinin (AHL21419.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0782