Learn More
Sino Biological Influenza A H5N8 (A/duck/Zhejiang/W24/2013) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40387V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Zhejiang/W24/2013(H5N8)) hemagglutinin (AHM24057.1)(Met1-Lys341), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0792