Learn More
Sino Biological Influenza B (B/Victoria/504/2000) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40391V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza B (B/Victoria/504/2000) Hemagglutinin (ABL77101.1) (Met1-Arg361), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0800