Learn More
Sino Biological Influenza A H1N1 (A/NewJersey/8/1976) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40392V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/NewJersey/8/1976(H1N1)) hemagglutinin (ACU80014.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0802