Learn More
Sino Biological Influenza A H3N2 (A/Victoria/3/1975) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40396V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/3/1975(H3N2)) hemagglutinin (AFM71967.1) (Met1-Arg346), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0814