Learn More
Sino Biological Influenza A H6N1 (A/quail/Hong Kong/1721-30/99) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40397V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/quail/Hong Kong/1721-30/99 (H6N1)) hemagglutinin (CAC84981.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0816