Learn More
Sino Biological HIV-1 (group M, subtype B, strain SHIV-89.6P) gp120 Protein (His Tag)
Supplier:  Sino Biological 40402V08H100
Encompass_Preferred
A DNA sequence encoding the HIV1(group M, subtype B, strain SHIV-89.6P) Envelope glycoprotein gp160 Protein(gp120 subunit) (AAC57427.1) (Thr31-Arg509) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0824