Learn More
Sino Biological HIV-1 (group M, subtype A, isolate 92RW020) Envelope glycoprotein gp160 Protein (gp120 subunit) (His Tag)
Supplier:  Sino Biological 40403V08H100
Encompass_Preferred
A DNA sequence encoding the HIV-1 envelope glycoprotein gp160 (AAT67478.1) extracellular domain (Glu31-Arg494),termed as gp120, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0828