Learn More
Sino Biological Human immunodeficiency virus 1 (HIV-1) (group M, subtype B, Isolate MN) gp120 Protein (His Tag)
Supplier:  Sino Biological 40405V08H100
Encompass_Preferred
A DNA sequence encoding the HIV1(group M, subtype B, isolate MN) Envelope glycoprotein gp160 Protein(gp120 subunit) (AAC31819.1) (Thr30-Arg513) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0836