Learn More
Sino Biological Simian immunodeficiency virus (SIV) (isolate F236) envelope glycoprotein gp120 Protein (His Tag)
Supplier:  Sino Biological 40415V08H100
Encompass_Preferred
A DNA sequence encoding the SIV envelope glycoprotein gp160 (CAA32487.1) extracellular domain (Gln24-Arg531),termed as gp120, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0848