Learn More
Sino Biological Rift Valley fever virus (TAN/Dod-002/07) RVFV-L Protein (His Tag)
Supplier:  Sino Biological 40423V08E100
Encompass_Preferred
A DNA sequence encoding the Rift Valley fever virus(TAN/Dod-002/07) L (AEB20483.1) (Met1-Phe250) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0852