Learn More
Aviva Systems Biology Corp SEPT6 Antibody - N-terminal region (ARP46217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPT6(septin 6). It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SEPT6(septin 6). Alternate names: RP5-876A24.2, KIAA0128, MGC16619, MGC20339, SEP2, SEPT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59273