Learn More
Aviva Systems Biology Corp SEPT10 Antibody - C-terminal region (ARP48438_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48438_T100
Encompass_Preferred
Rabbit polyclonal antibody recognizes the C terminal of SEPT10(septin 10) protein. It is suitable for WB application; 100ug/100ul
Catalog No. 50-014-59825