Learn More
Sino Biological Influenza B (B/Brisbane/3/2007) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40431V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Brisbane/3/2007) hemagglutinin (translated amino acids of EPI483156) (Met1-Arg361), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0857