Learn More
Aviva Systems Biology Corp SEPT11 Antibody - N-terminal region (ARP57173_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57173_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPT11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SEPT11(septin 11). Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62099