Learn More
Sino Biological Influenza A H10N7 (A/blue-winged teal/Louisiana/Sg-00073/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40433V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/blue-winged teal/Louisiana/Sg-00073/2007(H10N7)) hemagglutinin (ADH96304.1) (Met1-Val525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0861