Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Glycoprotein / GP-RBD Protein (Fc Tag)
Supplier:  Sino Biological 40442V02H100
Encompass_Preferred
A DNA sequence encoding the Zaire ebolavirus(strain H.sapiens-wt/GIN/2014/Kissidougou-C15) GP (AHX24649.1) (Met1-Phe308) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0873