Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Makona-Kissidougou-C15) GP / Glycoprotein Protein (His Tag)
Supplier:  Sino Biological 40442V08H2100
Encompass_Preferred
A DNA sequence encoding the Zaire ebolavirus (strain H.sapiens-wt/GIN/2014/Makona-Kissidougou-C15) GP (QDA39862.1) (Met1-Gln650) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0885