Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Glycoprotein / GP (Virion spike glycoprotein) Protein (His Tag)
Supplier:  Sino Biological 40442V08H4100
Encompass_Preferred
A DNA sequence encoding the Zaire ebolavirus(strain H.sapiens-wt/GIN/2014/Kissidougou-C15) GP (AHX24649.1) (Met1-Gln650, with the deletion of Thr309-Asn484) was expressed with a trimerization motif and a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0887