Learn More
Sino Biological Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40 / Matrix protein VP40 Protein (His Tag)
Catalog No. 501620903
Encompass_Preferred
A DNA sequence encoding the Bundibugyo ebolavirus(strain Uganda 2007) VP40 (YP_003815434.1) (Ala31-Lys326) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0903 Supplier Sino Biological Supplier No. 40448V07E100
A DNA sequence encoding the Bundibugyo ebolavirus(strain Uganda 2007) VP40 (YP_003815434.1) (Ala31-Lys326) was expressed with a polyhistidine tag at the N-terminus.