Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 Protein (His Tag)
Supplier:  Sino Biological 40454V07E100
Encompass_Preferred
A DNA sequence encoding the Zaire ebolavirus(strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 (AHX24653.1) (Met1-Gly233) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0905