Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/SLE/2014/ManoRiver-G3686.1) EBOV-G / Glycoprotein Protein (His Tag)
Supplier:  Sino Biological 40459V08H1100
Encompass_Preferred
A DNA sequence encoding the Zaire ebolavirus(strain Ebola virus H.sapiens-wt/SLE/2014/ManoRiver-G3686.1) GP (AIE11917.1) (Met1-Gln650,Ser46Asn,Phe291Arg) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0913