Learn More
Sino Biological Influenza B (B/Utah/02/2012) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40463V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Utah/02/2012) hemagglutinin (AFT64423.1) (Met1-Arg361), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0924