Learn More
Sino Biological Influenza A H5N6 (A/duck/Guangdong/GD01/2014) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40465V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Guangdong/GD01/2014 (H5N6)) hemagglutinin (AIA24403.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0932