Learn More
Sino Biological Enterovirus D68 (EV-D68) (strain Fermon) VP1 Protein (Fc Tag)
Supplier:  Sino Biological 40467V02B100
Encompass_Preferred
A DNA sequence encoding the Enterovirus D68 (EV-D68) (strain Fermon) VP1 (AAR98503.1) (Ser553-Thr861) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0938