Learn More
Sino Biological Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein (ECD, His Tag)
Supplier:  Sino Biological 40471V08B100
Encompass_Preferred
A DNA sequence encoding the envelope protein of Dengue virus (DENV)(type 2, strain New Guinea C/PUO-218 hybrid)(AAC59274.1) (Phe247-Gly675, with two amino acids "MV" at the N-terminal) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0942